Methyl-6-aminonicotinate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester
H27575-06
H27575-03
AAH27575-06EA
90.7
USD
AAH27575-06
AAH27575-03
Methyl-6-aminonicotinate 97%
Methyl-6-aminonicotinate
Purity: 97%
CAS No.: 36052-24-1
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: 158-161°
MDL No.: MFCD00797844
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Melting Pt: 158…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00797844 CAS Number: 36052-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...