3-Ethyl-5-methylisoxazole-4-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H27574-03 H27574-09
AAH27574-03EA 26.8 USD
AAH27574-03 AAH27574-09
3-Ethyl-5-methylisoxazole-4-carboxylic acid 97%
3-Ethyl-5-methylisoxazole-4-carboxylic acid

Purity: 97% CAS No.: 17147-85-2 Molecular Formula: C7H9NO3 Formula Weight: 155.15 Melting Point: 123-125° MDL No.: MFCD03422293

Formula: C₇H₉NO₃
MW: 155.15 g/mol
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD03422293
CAS Number: 17147-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR