cis-4-Aminocyclohexanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H27567-03 H27567-06
AAH27567-03EA 62.33 USD
AAH27567-03 AAH27567-06
cis-4-Aminocyclohexanecarboxylic acid 97%
cis-4-Aminocyclohexanecarboxylic acid

Purity: 97% CAS No.: 3685-23-2 Molecular Formula: C7H13NO2 Formula Weight: 143.19 Melting Point: 299-301° MDL No.: MFCD00191730

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Melting Pt: 299…301 °C
Storage Temperature: Ambient
MDL Number: MFCD00191730
CAS Number: 3685-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR