4-Bromo-2-(5-isoxazolyl)phenol 97%

Supplier: Thermo Scientific Chemicals
Warning

H27557-03 H27557-06 H27557-14
AAH27557-03EA 25.1 USD
AAH27557-03 AAH27557-06 AAH27557-14
4-Bromo-2-(5-isoxazolyl)phenol 97%
4-Bromo-2-(5-isoxazolyl)phenol

Purity: 97% CAS No.: 213690-27-8 Molecular Formula: C9H6BrNO2 Formula Weight: 240.05 Melting Point: 207-212° MDL No.: MFCD00799493

Formula: C₉H₆BrNO₂
MW: 240.05 g/mol
Melting Pt: 207…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00799493
CAS Number: 213690-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR