4-n-Pentyloxyphenylacetylene 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Eth-1-ynyl-4-(pentyloxy)benzene

H27555-03 H27555-09
AAH27555-03EA 39.4 USD
AAH27555-03 AAH27555-09
4-n-Pentyloxyphenylacetylene 99%
4-n-Pentyloxyphenylacetylene

Purity: 99% CAS No.: 79887-16-4 Molecular Formula: C13H16O Formula Weight: 188.27 Flash Point: 110°(230°F) Refractive Index: 1.5310 Boiling Point: 113-115°/2mm MDL No.: MFCD00173877

Formula: C₁₃H₁₆O
MW: 188.27 g/mol
Boiling Pt: 113…115 °C/2 mm
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00173877
CAS Number: 79887-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR