Ethyl-trans-4-bromocinnamate 98%

Supplier: Thermo Scientific Chemicals
Warning

H27544-06 H27544-14
AAH27544-06EA 72.6 USD
AAH27544-06 AAH27544-14
Ethyl-trans-4-bromocinnamate 98%
Ethyl-trans-4-bromocinnamate

Purity: 98% CAS No.: 24393-53-1 Molecular Formula: C11H11BrO2 Formula Weight: 255.11 Flash Point: 110°(230°F) Refractive Index: 1.5970 Boiling Point: 180°/18mm MDL No.: MFCD00012226

Formula: C₁₁H₁₁BrO₂
Boiling Pt: 180 °C/18 mm
Density: 1.359
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012226
CAS Number: 24393-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR