(S)-(+)-3-Fluoropyrrolidine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-3-Fluoropyrrolidine hydrochloride
H27536-MD
H27536-03
AAH27536-MDEA
51.9
USD
AAH27536-MD
AAH27536-03
AAH27536-06
(S)-(+)-3-Fluoropyrrolidine hydrochloride 97%
(S)-(+)-3-Fluoropyrrolidine hydrochloride
Purity: 97%
CAS No.: 136725-53-6
Molecular Formula: C4H8FN.HCl
Formula Weight: 125.57
Melting Point: 183-186°
MDL No.: MFCD04038718
Formula:
C₄H₉ClFN MW: 125.57 g/mol Melting Pt: 183…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038718 CAS Number: 136725-53-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...