5-(3-Hydroxyphenyl)-1H-tetrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(1H-Tetrazol-5-yl)phenol

H27529-03 H27529-06 H27529-MD
AAH27529-03EA 150 USD
AAH27529-03 AAH27529-06 AAH27529-MD
5-(3-Hydroxyphenyl)-1H-tetrazole 97%
5-(3-Hydroxyphenyl)-1H-tetrazole

Purity: 97% CAS No.: 96859-34-6 Molecular Formula: C7H6N4O Formula Weight: 162.16 Melting Point: ca 220° dec. MDL No.: MFCD06797247

Formula: C₇H₆N₄O
MW: 162.16 g/mol
Melting Pt: ca 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06797247
CAS Number: 96859-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR