1-Phenylcyclopropanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Phenyl-1-cyclopropanecarboxylic acid

H27525-14 H27525-06
AAH27525-14EA 171 USD
AAH27525-14 AAH27525-06
1-Phenylcyclopropanecarboxylic acid 97%
1-Phenylcyclopropanecarboxylic acid

Purity: 97% CAS No.: 6120-95-2 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 85-89° MDL No.: MFCD00001288

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00001288
CAS Number: 6120-95-2
EINECS: 228-090-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR