5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Amino-5-(4-pyridinyl)-1,3,4-thiadiazole, 5-(4-Pyridyl)-1,3,4-thiadiazol-2-amine, 2-Amino-5-(4-pyridyl)-1,3,4-thiadiazole
H27514-03
H27514-09
AAH27514-03EA
20.2
USD
AAH27514-03
AAH27514-09
5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine 97%
5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine
Purity: 97%
CAS No.: 2002-04-2
Molecular Formula: C7H6N4S
Formula Weight: 178.21
Melting Point: 242-248°
MDL No.: MFCD00473692
Formula:
C₇H₆N₄S MW: 178.22 g/mol Melting Pt: 242…248 °C Storage Temperature: Ambient |
MDL Number:
MFCD00473692 CAS Number: 2002-04-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...