2-Cyclopentene-1-acetic acid 90% remainder mainly 3-cyclopentene-1-acetic acid, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27507-06
H27507-14
AAH27507-06EA
74.4
USD
AAH27507-06
AAH27507-14
2-Cyclopentene-1-acetic acid 90% remainder mainly 3-cyclopentene-1-acetic acid, tech.
2-Cyclopentene-1-acetic acid
Purity: 90%
CAS No.: 13668-61-6
Molecular Formula: C7H10O2
Formula Weight: 126.15
Flash Point: 143°(289°F)
Refractive Index: 1.4680
Odor: Rancid
Melting Point: 19°
Boiling Point: 238°
Beilstein Reference: 2042172
MDL No.: MFCD00001400
Formula:
C₇H₁₀O₂ Boiling Pt: 238 °C Melting Pt: 19 °C Density: 1.047 Flash Pt: 143 °C(289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001400 CAS Number: 13668-61-6 EINECS: 237-146-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...