1-Cyclopropyl-4-piperidinone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Cyclopropylpiperidin-4-one

H27495-03 H27495-06 H27495-14
AAH27495-03EA 59.1 USD
AAH27495-03 AAH27495-06 AAH27495-14
1-Cyclopropyl-4-piperidinone 98%
1-Cyclopropyl-4-piperidinone

Purity: 98% CAS No.: 62813-01-8 Molecular Formula: C8H13NO Formula Weight: 139.20 MDL No.: MFCD07374375

Formula: C₈H₁₃NO
MW: 139.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374375
CAS Number: 62813-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR