6-Bromo-2H-1,4-benzoxazin-3(4H)-one 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27493-09
H27493-03
AAH27493-09EA
261
USD
AAH27493-09
AAH27493-03
6-Bromo-2H-1,4-benzoxazin-3(4H)-one 95%
6-Bromo-2H-1,4-benzoxazin-3(4H)-one
Purity: 95%
CAS No.: 24036-52-0
Molecular Formula: C8H6BrNO2
Formula Weight: 228.04
Melting Point: 220-225°
MDL No.: MFCD00461173
Formula:
C₈H₆BrNO₂ Melting Pt: 220…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00461173 CAS Number: 24036-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...