trans-4-Aminocyclohexanecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
H27489-06
H27489-03
AAH27489-06EA
624
USD
AAH27489-06
AAH27489-03
trans-4-Aminocyclohexanecarboxylic acid 97%
trans-4-Aminocyclohexanecarboxylic acid
Purity: 97%
CAS No.: 3685-25-4
Molecular Formula: C7H13NO2
Formula Weight: 143.19
Melting Point:
Formula:
C₇H₁₃NO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00209953 CAS Number: 3685-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...