Methyl-3-aminopropanoate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
beta-Alanine methyl ester hydrochloride, H-β-Ala-OMe·HCl, β-Alanine methyl ester hydrochloride, 3-Aminopropionic acid Methyl Ester Hydrochloride, 3-Aminopropionic Acid Methyl Ester Hydrochloride, Methyl 3-aminopropanoate hydrochloride, Methyl 3-Aminopropionate Hydrochloride, H-β-Ala-OMe.HCl
H27483-14
H27483-06
AAH27483-14EA
184
USD
AAH27483-14
AAH27483-06
Methyl-3-aminopropanoate hydrochloride 98%
Methyl-3-aminopropanoate hydrochloride
Purity: 98%
CAS No.: 3196-73-4
Molecular Formula: C4H9NO2.HCl
Formula Weight: 139.58
Storage Temperature: 2°C to 8°C
Melting Point: 102-104°
Beilstein Reference: 3556748
MDL No.: MFCD00039060
Formula:
C₄H₉NO₂·HCl MW: 139.6 g/mol Melting Pt: 102…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039060 CAS Number: 3196-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...