4-Aminophenylboronic acid hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Aminobenzeneboronic acid hydrochloride, (4-Aminophenyl)boronic acid hydrochloride

H27479-ME H27479-MC
AAH27479-MEEA 243 USD
AAH27479-ME AAH27479-MC
4-Aminophenylboronic acid hydrochloride 97%
4-Aminophenylboronic acid hydrochloride

Purity: 97% CAS No.: 80460-73-7 Molecular Formula: C6H8BNO2.HCl Formula Weight: 173.41 Melting Point: 125-132° MDL No.: MFCD03001333

Formula: C₆H₉BClNO₂
MW: 173.41 g/mol
Melting Pt: 125…132 °C
Storage Temperature: Ambient
MDL Number: MFCD03001333
CAS Number: 80460-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR