4-(Aminomethyl)benzonitrile hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Aminomethyl)benzonitrile HCl, α-Amino-p-tolunitrile Hydrochloride, 4-(Aminomethyl)benzonitrile Hydrochloride, 4-Cyanobenzylamine Hydrochloride
H27473-06
H27473-14
AAH27473-06EA
78
USD
AAH27473-06
AAH27473-14
4-(Aminomethyl)benzonitrile hydrochloride 97%
4-(Aminomethyl)benzonitrile hydrochloride
Purity: 97%
CAS No.: 15996-76-6
Molecular Formula: C8H8N2.HCl
Formula Weight: 168.62
Melting Point: 279-284°
MDL No.: MFCD01861472
Formula:
C₈H₉ClN₂ MW: 168.62 g/mol Melting Pt: 279…284 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861472 CAS Number: 15996-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...