2-(Benzylamino)-5-pyridineboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-(Benzylamino)pyridine-3-boronic acid pinacol ester, N-Benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

H27463-03 H27463-MD
AAH27463-03EA 224 USD
AAH27463-03 AAH27463-MD
2-(Benzylamino)-5-pyridineboronic acid pinacol ester 95%
2-(Benzylamino)-5-pyridineboronic acid pinacol ester

Purity: 95% CAS No.: 1073354-27-4 Molecular Formula: C18H23BN2O2 Formula Weight: 310.20 Melting Point: 135-138° MDL No.: MFCD06798270

Formula: C₁₈H₂₃BN₂O₂
Melting Pt: 135…138 °C
Storage Temperature: Ambient
MDL Number: MFCD06798270
CAS Number: 1073354-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR