2-Chloro-5-fluoropyridine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H27462-03
H27462-06
AAH27462-03EA
35
USD
AAH27462-03
AAH27462-06
2-Chloro-5-fluoropyridine 95%
2-Chloro-5-fluoropyridine
Purity: 95%
CAS No.: 31301-51-6
Molecular Formula: C5H3ClFN
Formula Weight: 131.54
Flash Point: 42°(108°F)
Refractive Index: 1.503
Melting Point: 19°
Boiling Point: 74°/38mm
UN No.: 2929
MDL No.: MFCD03095332
Formula:
C₅H₃ClFN Boiling Pt: 74 °C/38 mm Melting Pt: 19 °C Density: 1.297 Flash Pt: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03095332 CAS Number: 31301-51-6 UN: 1993 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...