3-Nitro-o-phenylenediamine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Nitrobenzene-1,2-diamine, 3-Nitro-1,2-phenylenediamine
B20201-14
B20201-03
B20201-06
AAB20201-14EA
766
USD
AAB20201-14
AAAB20201-03
AAAB20201-06
3-Nitro-o-phenylenediamine 98%
3-Nitro-o-phenylenediamine
Purity: 98%
CAS No.: 3694-52-8
Molecular Formula: C6H7N3O2
Formula Weight: 153.14
Melting Point: 157-162°
Beilstein Reference: 909344
Formula:
C₆H₇N₃O₂ MW: 153.14 g/mol Melting Pt: 157…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007722 CAS Number: 3694-52-8 EINECS: 223-013-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...