Copper(I) oxide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cuprous oxide, Dicopper oxide

A14436-0E A14436-36
AAA14436-22EA 25.31 USD
AAA14436-22 AAAA14436-0E AAAA14436-36
Copper(I) oxide 97%
Copper(I) oxide

Purity: 97% CAS No.: 1317-39-1 Molecular Formula: Cu2O Formula Weight: 143.08 Odor: Odorless Melting Point: 1235° Boiling Point: 1800° dec. Merck Reference: 14,2664 UN No.: 3077 MDL No.: MFCD00010974

Formula: Cu₂O
MW: 143.09 g/mol
Boiling Pt: 1800 °C (1013 hPa)
Melting Pt: 1232 °C
Density: 5.7 to 6.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010974
CAS Number: 1317-39-1
EINECS: 215-270-7
UN: 3077
ADR: 9,III
Merck Index: 13,02694

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR