Triethyloxonium tetrafluoroborate 1.0 M in dichloromethane
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11678-AC
A11678-AP
A11678-AE
AAAA11678-ACEA
43.3
USD
AAAA11678-AC
AAAA11678-AP
AAAA11678-AE
Triethyloxonium tetrafluoroborate 1.0 M in dichloromethane
Triethyloxonium tetrafluoroborate
CAS No.: 368-39-8
Molecular Formula: C6H15BF4O
Formula Weight: 189.99
Storage Temperature: -30°C to -10°C
Odor: Chloroform-like
Beilstein Reference: 3598090
UN No.: 2922
MDL No.: MFCD00044423
Formula:
C₆H₁₅BF₄O MW: 189.99 g/mol Density: 1.328 Storage Temperature: Refrigerator |
MDL Number:
MFCD00044423 CAS Number: 368-39-8 EINECS: 206-705-1 UN: 2922 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...