Polyethylene glycol dimethacrylate, M.W. 8,000

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: PEGDMA, Polyethylene Glycol Dimethacrylate average Mn 550, PEG dimethacrylate, Polyethylene Glycol Dimethacrylate

046985-03 046985-06
AA46985-03EA 107 USD
AA46985-03 AA46985-06 AA46985-MD
Polyethylene glycol dimethacrylate, M.W. 8,000
Polyethylene glycol dimethacrylate

Granular solid


CAS No.: 25852-47-5 Molecular Formula: CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3 Storage Temperature: -30°C to -10°C Physical Form: Granular solid Sensitivity: Moisture and Light Sensitive Appearance: White to off-white MDL No.: MFCD00081877

Formula: (C₂H₄O)nC₈H₁₀O₃
Storage Temperature: Freezer
MDL Number: MFCD00014932
CAS Number: 25852-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR