Methoxypolyethylene glycol amine, M.W. 5,000
Supplier: Thermo Scientific Chemicals
046924-03
046924-06
046924-MD
AA46924-03EA
177.83
USD
AA46924-03
AA46924-06
AA46924-MD
Methoxypolyethylene glycol amine, M.W. 5,000
Methoxypolyethylene glycol amine
Granular solid
CAS No.: 80506-64-5
Molecular Formula: H2NCH2CH2(OCH2CH2)nOCH3
Physical Form: Granular solid
Appearance: White to off-white
MDL No.: MFCD00131599
Formula:
H₂NCH₂CH₂(OCH₂CH₂)nOCH₃ Storage Temperature: Ambient |
MDL Number:
MFCD00131599 CAS Number: 80506-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...