Polyoxyethylene bis(amine), M.W. 8,000
Supplier: Thermo Scientific Chemicals
Synonyms:
PEG-bis-amine, Amine-PEG-Amine, O,O′-Bis(2-aminoethyl)polyethylene glycol, PEG-diamine, Amine-PEG-amine (MW 4000), Poly(ethylene glycol) diamine, Polyoxyethylene bis(amine), Diaminopolyethylene glycol
046903-03
046903-06
046903-MD
AA46903-03EA
117
USD
AA46903-03
AA46903-06
AA46903-MD
Polyoxyethylene bis(amine), M.W. 8,000
Polyoxyethylene bis(amine)
Granular solid
CAS No.: 24991-53-5
Molecular Formula: H2NCH2CH2(OCH2CH2)nNH2
Physical Form: Granular solid
Sensitivity: Air Sensitive
Appearance: White to off-white
MDL No.: MFCD00132004
Formula:
H₂NCH₂CH₂(OCH₂CH₂)nNH₂ Storage Temperature: Ambient |
MDL Number:
MFCD00132004 CAS Number: 24991-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...