Methoxypolyethylene glycol amine, M.W. 1,000
Supplier: Thermo Scientific Chemicals
Synonyms:
Amino polyethylene glycol monomethyl ether-average M.W. 2000, m-PEG12-amine, mPEG-amine, Amino polyethylene glycol monomethyl ether, mPEG-NH2
046643-03
046643-06
046643-MD
AA46643-03EA
136
USD
AA46643-03
AA46643-06
AA46643-MD
Methoxypolyethylene glycol amine, M.W. 1,000
Methoxypolyethylene glycol amine
Granular solid
CAS No.: 80506-64-5
Molecular Formula: H2NCH2CH2(OCH2CH2)nOCH3
Physical Form: Granular solid
Appearance: White to off-white
MDL No.: MFCD00131599
Formula:
H₂NCH₂CH₂(OCH₂CH₂)nOCH₃ Storage Temperature: Ambient |
MDL Number:
MFCD00131599 CAS Number: 80506-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...