Polyethylene glycol diacrylate (PEGDA), M.W. 1,000

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: PEG hydrogel, Polyethylene glycol diacrylate, Poly(ethylene glycol) diacrylate, PEG diacrylate, PEGDA, PEG bisacrylate

046493-03 046493-06 046493-MD
AA46493-03EA 131 USD
AA46493-03 AA46493-06 AA46493-MD
Polyethylene glycol diacrylate (PEGDA), M.W. 1,000
Polyethylene glycol diacrylate (PEGDA)

Granular solid


CAS No.: 26570-48-9 Molecular Formula: CH2=COC(OCH2CH2)nOCOC=CH2 Storage Temperature: 2°C to 8°C Physical Form: Granular solid Appearance: White to off-white MDL No.: MFCD00081876

Formula: CH₂=COC(OCH₂CH₂)nOCOC=CH₂
Storage Temperature: Refrigerator
MDL Number: MFCD00081876
CAS Number: 26570-48-9
EINECS: 607-960-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR