Polyethylene glycol monomethyl ether methanesulfonate, M.W. 1,000

Supplier: Thermo Scientific Chemicals
Warning

046437-03 046437-06
AA46437-03EA 92.8 USD
AA46437-03 AA46437-06 AA46437-MD
Polyethylene glycol monomethyl ether methanesulfonate, M.W. 1,000
Polyethylene glycol monomethyl ether methanesulfonate

Granular solid


CAS No.: 175172-61-9 Molecular Formula: CH3(OCH2CH2)nOSO2CH3 Physical Form: Granular solid Appearance: White to off-white MDL No.: MFCD03100948

Formula: CH₃(OCH₂CH₂)nOSO₂CH₃
Storage Temperature: Ambient
MDL Number: MFCD03100948
CAS Number: 175172-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR