Methoxypolyethylene glycol amine, M.W. 13,000
Supplier: Thermo Scientific Chemicals
Synonyms:
Amino polyethylene glycol monomethyl ether-average M.W. 2000, m-PEG12-amine, mPEG-amine, Amino polyethylene glycol monomethyl ether, mPEG-NH2
046433-03
046433-06
046433-MD
AA46433-03EA
169
USD
AA46433-03
AA46433-06
AA46433-MD
Methoxypolyethylene glycol amine, M.W. 13,000
Methoxypolyethylene glycol amine
Granular solid
CAS No.: 80506-64-5
Molecular Formula: H2NCH2CH2(OCH2CH2)nOCH3
Physical Form: Granular solid
Appearance: White to off-white
MDL No.: MFCD00131599
Formula:
H₂NCH₂CH₂(OCH₂CH₂)nOCH₃ Storage Temperature: Ambient |
MDL Number:
MFCD00131599 CAS Number: 80506-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...