Polyethylene glycol methyl ether acrylate, M.W. 5,000
Supplier: Thermo Scientific Chemicals
Synonyms:
mPEG-acrylate, Polyethylene glycol monomethyl ether acrylate
046388-03
046388-06
AA46388-03EA
104
USD
AA46388-03
AA46388-06
AA46388-MD
Polyethylene glycol methyl ether acrylate, M.W. 5,000
Polyethylene glycol methyl ether acrylate
Granular solid
CAS No.: 32171-39-4
Molecular Formula: CH3(OCH2CH2)nOCOC=CH2
Storage Temperature: 2°C to 8°C
Physical Form: Granular solid
Appearance: White to off-white
MDL No.: MFCD00803685
Formula:
CH₃(OCH₂CH₂)nOCOC=CH₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00803685 CAS Number: 32171-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...