Nitinol (alloy M), tension coil spring wire diameter 0.75 mm (0.030 in), mean coil diameter 5.55 mm (0.22 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
045655-KM
045655-KJ
045655-KN
AA45655-KMEA
490
USD
AA45655-KM
AA45655-KJ
AA45655-KN
Nitinol (alloy M), tension coil spring wire diameter 0.75 mm (0.030 in), mean coil diameter 5.55 mm (0.22 in)
Nitinol (alloy M)
Odor: Odorless
MDL No.: MFCD02091734
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...