Copper, alloy 101, plate, Oxygen-Free High Conductivity (OFHC), Thickness 2.4 mm (0.093 in)

Supplier: Thermo Scientific Chemicals

045210-HB 045210-GT
AA45210-HBEA 443 USD
AA45210-HB AA45210-GT
Copper, alloy 101, plate, Oxygen-Free High Conductivity (OFHC), Thickness 2.4 mm (0.093 in)
Copper, alloy 101

MDL No.: MFCD00010965

Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR