Palladium 5% on activated carbon paste (50% water) 5R58

Supplier: Thermo Scientific Chemicals

045132-06 045132-22 045132-14
AA45132-06EA 82.7 USD
AA45132-06 AA45132-22 AA45132-14
Palladium 5% on activated carbon paste (50% water) 5R58
Palladium

CAS No.: 7440-05-3 Appearance: Black Odor: Odorless MDL No.: MFCD03457879

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR