Nickel iron molybdenum, foil, annealed, Thickness: 0.102 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
045092-HB
045092-4K
AA45092-HBEA
100
USD
AA45092-HB
AA45092-4K
Nickel iron molybdenum, foil, annealed, Thickness: 0.102 mm (0.004 in)
Nickel iron molybdenum
ca. 40.1 g/150 mm x 300 mm
Molecular Formula: Ni:Fe:Mo; 80:15:4.2 wt%
Physical Form: ≈40.1g/150mmx300mm
MDL No.: MFCD09910537
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...