Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness 0.38 mm (0.015 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
044954-FI
044954-FF
AA44954-FIEA
247
USD
AA44954-FI
AA44954-FF
Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness 0.38 mm (0.015 in)
Nitinol
ca 1.56g/25x25mm; Af approx. 10ºC
Physical Form: ≈1.56g/25x25mm; Af approx. 10°C
Odor: Odorless
MDL No.: MFCD02091734
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...