Antimony tin oxide ≥99.5%, NanoArc®

Supplier: Thermo Scientific Chemicals
Warning

044930-14 044930-22 044930-36
AA44930-14EA 35.6 USD
AA44930-14 AA44930-22 AA44930-36
Antimony tin oxide ≥99.5%, NanoArc®
Antimony tin oxide

22-44nm APS Powder, S.A. 20-40m2/g


Applications: Moderately conductive particle. The incorporation of NanoTek™ antimony tin oxide products in coating systems, films or composites may help in reducing the buildup of static charge


Applications: Moderately conductive particle. The incorporation of NanoTek™ antimony tin oxide products in coating systems, films or composites may help in reducing the buildup of static charge

Formula: Sb₂O₅:SnO₂₁₅:₈₅%
Storage Temperature: Ambient
MDL Number: MFCD00799153
UN: 1549
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR