Iron(III) isopropoxide ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Iron(III) i-propoxide
Ampuled under argon
044873-03
044873-14
044873-06
AAA44873-03EA
70.1
USD
AAA44873-03
AAA44873-14
AAA44873-06
Iron(III) isopropoxide ≥98%
Iron(III) isopropoxide
Solid mass, Ampouled under argon
Purity: 98%
CAS No.: 14995-22-3
Molecular Formula: C9H21FeO3
Formula Weight: 233.11
Physical Form: Solid mass, Ampouled under argon
Sensitivity: Moisture Sensitive
Appearance: Dark grey
UN No.: 3181
MDL No.: MFCD00070436
Formula:
C₉H₂₁FeO₃ MW: 233.109 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070436 CAS Number: 14995-22-3 UN: 3181 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...