Poly(3-octylthiophene-2,5-diyl) regioregular low metals

Supplier: Thermo Scientific Chemicals

044777-03 044777-01
AAA44777-03EA 741 USD
AAA44777-03 AAA44777-01
Poly(3-octylthiophene-2,5-diyl) regioregular low metals
Poly(3-octylthiophene-2,5-diyl)

Powder


Applications: Conducting polymers


Applications: Conducting polymers

Storage Temperature: Ambient
MDL Number: MFCD00241412
CAS Number: 104934-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR