Potassium hexachlororuthenate(III) (≥23% Ru) ≥99.99% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Warning

044547-06 044547-03
AAA44547-06EA 242 USD
AAA44547-06 AAA44547-03
Potassium hexachlororuthenate(III) (≥23% Ru) ≥99.99% (metals basis), Premion®
Potassium hexachlororuthenate(III)

Crystalline


Purity: 99.99% CAS No.: 25443-63-4 Molecular Formula: K3RuCl6 Formula Weight: 431.09 Physical Form: Crystalline MDL No.: MFCD00011390

Formula: K₃RuCl₆
MW: 431.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011390
CAS Number: 25443-63-4
EINECS: 246-983-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR