Poly(styrene-divinylbenzene), aminomethylated, cross-linked with 1% DVB 100-200 mesh
Supplier: Thermo Scientific Chemicals
044392-14
044392-06
AAA44392-14EA
209
USD
AAA44392-14
AAA44392-22
AAA44392-06
Poly(styrene-divinylbenzene), aminomethylated, cross-linked with 1% DVB 100-200 mesh
Poly(styrene-divinylbenzene), aminomethylated
Powder
Applications: Formation of amide groups using Boc strategy; as a scavenger resin
Applications: Formation of amide groups using Boc strategy. As a scavenger resin
Storage Temperature:
Ambient |
MDL Number:
MFCD00146442 CAS Number: 89551-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...