Sodium dodecyl sulphate (SDS) solution_SC 10%, ultrapure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

J77504-K2 J77504-AE J77504-AP
101174-138EA 221.82 USD
101174-138 101171-450 101176-116
Sodium dodecyl sulphate (SDS) solution_SC 10%, ultrapure
Sodium dodecyl sulphate (SDS)

10% (w/v) solution of sodium dodecyl sulfate (SDS) in high purity dH2O.


  • A(280 nm, 3%, 1 cm): ≤ 0.1
  • A(230 nm, 3%, 1 cm): ≤ 0.2


Solution is 0.2 μm filtered and dispensed into durable, square Nalgene bottles.


Precaución: Note: Precipitation of SDS is not unusual. Warm gently to redissolve.

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR