PBS, 10X Solution, pH 7.4, Molecular Biology Grade, Ultrapure, Thermo Scientific

Supplier: Thermo Fisher Scientific

J75889-K8 J75889-AE
101175-844EA 230.54 USD
101175-844 101171-424 101175-842
PBS, 10X Solution, pH 7.4, Molecular Biology Grade, Ultrapure, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

10X PBS (Phosphate Buffered Saline) solution consisting of 80.6mM sodium phosphate, 19.4mM potassium phosphate, 27mM KCl and 1.37M NaCl in high purity dH2O, pH 7.4


  • Contaminant Testing:
  • DNase (endo): Passes test
  • DNase (exo): Passes test
  • RNase: Passes test
  • Protease: Passes test


Solution is 0.2 μm filtered and dispensed into durable, square Nalgene bottles. The 5L size is supplied in an upright carton with a convenient dispensing spout. Dilute to 1X with high purity dH2O for a solution consisting of 10mM phosphate, pH 7.4 ± 0.1, 2.7mM potassium chloride and 137mM NaCl.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR