Water, ultrapure Molecular_Biology RNase_free, DEPC_treated

Supplier: Thermo Fisher Scientific

Synonyms: Hydrogen oxide

J70783-AC J70783-AP J70783-AE J70783-K2
101170-290EA 44.65 USD
101170-290 101174-830 101175-822 101175-820
Water, ultrapure Molecular_Biology RNase_free, DEPC_treated
Water

High purity dH2O treated overnight with 0.1% Diethylpyrocarbonate. Product is 0.2 μm filtered and dispensed into vials and/or durable, square Nalgene bottles, then autoclaved.


Contaminant Testing:
RNase: Passes test
DNase (endo): Passes test
DNase (exo): Passes test
Enzyme Inhibition: Passes test

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1.000
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 14,10039

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR