Zirconium(IV) Propoxide ∼70% in 1-propanol
Supplier: TCI America
Synonyms:
Zirconium(IV) Tetrapropoxide, Zirconium(IV) n-propoxide
Z0017-100G
Z0017-25G
TCZ0017-100GEA
64
USD
TCZ0017-100G
TCZ0017-25G
Zirconium(IV) Propoxide ∼70% in 1-propanol
Zirconium(IV) Propoxide
CAS Number: 23519-77-9
MDL Number: MFCD00015307
Molecular Formula: C12H28O4Zr
Molecular Weight: 327.58
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): 28
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: Zirconium(IV) Tetrapropoxide
Formula:
C₁₂H₂₈O₄Zr MW: 327.58 g/mol Density: 1.04 (20 °C) Flash Pt: 28 °C |
MDL Number:
MFCD00015307 CAS Number: 23519-77-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...