Zirconium(IV) butoxide ∼80% in 1-butanol

Supplier: TCI America

Synonyms: Zirconium(IV) n-butoxide, Zirconium(IV) Tetrabutoxide

Z0016-500G Z0016-25G
TCZ0016-500GEA 148 USD
TCZ0016-500G TCZ0016-25G
Zirconium(IV) butoxide ∼80% in 1-butanol
Zirconium(IV) butoxide

CAS Number: 1071-76-7
MDL Number: MFCD00048765
Molecular Formula: C16H36O4Zr
Molecular Weight: 383.68
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): 38
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Zirconium(IV) Tetrabutoxide

Formula: C₁₆H₃₆O₄Zr
MW: 383.68 g/mol
Density: 1.06 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00048765
CAS Number: 1071-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4cb12459eea35f85fe6c21bd2aab91205a6373a&vid=a3c766c59bdb28d83271a026de33e3d819946ea8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR