Sodium vinylsulfonate 25% ∼2.3 mol/L in water

Supplier: TCI America

Synonyms: Vinylsulphonic acid Sodium Salt

V0043-500ML V0043-25ML
TCV0043-500MLEA 97.65 USD
TCV0043-500ML TCV0043-025ML
Sodium vinylsulfonate 25% ∼2.3 mol/L in water
Sodium vinylsulfonate

CAS Number: 3039-83-6
MDL Number: MFCD00007520
Molecular Formula: C2H4O3S
Molecular Weight: 130.09
Form: Clear Liquid
Color: Very Pale Yellow
Melting point (°C): -20


Other Product Names / Synonyms: Vinylsulfonic Acid Sodium Salt

Formula: C₂H₄O₃S
Melting Pt: –20 °C
MDL Number: MFCD00007520
CAS Number: 3039-83-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8814c65b624102535f57d106f04f372da346035&vid=ac392ed19e2d1dd0ee8e29fbc9b07db55a3264e9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR