4,4,4-Trifluorobutyric acid ≥98.0%
Supplier: TCI America
T2670-1G
T2670-5G
TCT2670-1GEA
24
USD
TCT2670-1G
TCT2670-5G
4,4,4-Trifluorobutyric acid ≥98.0%
4,4,4-Trifluorobutyric acid
CAS Number: 406-93-9
MDL Number: MFCD00077604
Molecular Formula: C4H5F3O2
Molecular Weight: 142.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 167
Melting point (°C): 31
Formula:
C₄H₅F₃O₂ MW: 142.08 g/mol Boiling Pt: 167 °C Melting Pt: 31 °C |
MDL Number:
MFCD00077604 CAS Number: 406-93-9 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...