2,4,6-Tribromoresorcinol ≥98.0%

Supplier: TCI America

T2620-1G T2620-5G
TCT2620-1GEA 36 USD
TCT2620-1G TCT2620-5G
2,4,6-Tribromoresorcinol ≥98.0%
2,4,6-Tribromoresorcinol

CAS Number: 2437-49-2
MDL Number: MFCD00009717
Molecular Formula: C6H3Br3O2
Molecular Weight: 346.80
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 114


Other Product Names / Synonyms: 1,3,5-Tribromo-2,4-dihydroxybenzene

Formula: C₆H₃Br₃O₂
Melting Pt: 114 °C
MDL Number: MFCD00009717
CAS Number: 2437-49-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5b69c3584e849f3dc65e43d23f0482878092f4a&vid=e5b69c3584e849f3dc65e43d23f0482878092f4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR