2,4,5-Trimethyloxazole ≥98.0%

Supplier: TCI America

T2595-5G
TCT2595-5GEA 41 USD
TCT2595-5G
2,4,5-Trimethyloxazole ≥98.0%
2,4,5-Trimethyloxazole

CAS Number: 20662-84-4
MDL Number: MFCD00005308
Molecular Formula: C6H9NO
Molecular Weight: 111.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 134
Flash Point (°C): 33
Specific Gravity (20/20): 0.96

Formula: C₆H₉NO
MW: 111.14 g/mol
Boiling Pt: 134 °C
Density: 0.96 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00005308
CAS Number: 20662-84-4
https://pr.vwr.com/stibo/web/std.lang.all/63/07/28996307.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR