2-(Trifluoromethyl)thiophenol ≥95.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(Trifluoromethyl)benzene-1-thiol, 2-(Trifluoromethyl)benzenethiol, 2-Mercaptobenzotrifluoride
T2519-1G
T2519-5G
TCT2519-1GEA
42
USD
TCT2519-1G
TCT2519-5G
2-(Trifluoromethyl)thiophenol ≥95.0% (by GC, titration analysis)
2-(Trifluoromethyl)thiophenol
CAS Number: 13333-97-6
MDL Number: MFCD00068896
Molecular Formula: C7H5F3S
Molecular Weight: 178.17
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 64
Flash Point (°C): 68
Specific Gravity (20/20): 1.34
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(Trifluoromethyl)thiophenol, 2-Mercaptobenzotrifluoride
Formula:
C₇H₅F₃S MW: 178.18 g/mol Boiling Pt: 64 °C Density: 1.34 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00068896 CAS Number: 13333-97-6 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >95.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...